IDO Monoclonal Antibody (eyedio), eFluor™ 660, eBioscience™ - FAQs
IDO Monoclonal Antibody (eyedio), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IDO Monoclonal Antibody (eyedio), eFluor™ 660, eBioscience™ - FAQs (50-9477-42, 50-9477-41)