Marginal Zone B Cells Monoclonal Antibody (HIS57), eFluor™ 660, eBioscience™ - FAQs
Marginal Zone B Cells Monoclonal Antibody (HIS57), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Marginal Zone B Cells Monoclonal Antibody (HIS57), eFluor™ 660, eBioscience™ - FAQs (50-9489-80)