TIGIT Monoclonal Antibody (GIGD7), eFluor™ 660, eBioscience™ - FAQs
TIGIT Monoclonal Antibody (GIGD7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for TIGIT Monoclonal Antibody (GIGD7), eFluor™ 660, eBioscience™ - FAQs (50-9501-82, 50-9501-80)