Musashi-2 Monoclonal Antibody (C1), eFluor™ 660, eBioscience™ - FAQs
Musashi-2 Monoclonal Antibody (C1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Musashi-2 Monoclonal Antibody (C1), eFluor™ 660, eBioscience™ - FAQs (50-9677-80, 50-9677-82)