Cyclin E Monoclonal Antibody (HE12), eFluor™ 660, eBioscience™ - FAQs
Cyclin E Monoclonal Antibody (HE12), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Cyclin E Monoclonal Antibody (HE12), eFluor™ 660, eBioscience™ - FAQs (50-9714-80)