S100A8 Monoclonal Antibody (CF-145), eFluor™ 660, eBioscience™ - FAQs
S100A8 Monoclonal Antibody (CF-145), eFluor™ 660, eBioscience™ - FAQs
View additional product information for S100A8 Monoclonal Antibody (CF-145), eFluor™ 660, eBioscience™ - FAQs (50-9745-41, 50-9745-42)