Desmin Monoclonal Antibody (DE-U-10), eFluor™ 660, eBioscience™ - FAQs
Desmin Monoclonal Antibody (DE-U-10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Desmin Monoclonal Antibody (DE-U-10), eFluor™ 660, eBioscience™ - FAQs (50-9747-80, 50-9747-82)