IDO Monoclonal Antibody (V1NC3IDO), eFluor™ 660, eBioscience™ - FAQs
IDO Monoclonal Antibody (V1NC3IDO), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IDO Monoclonal Antibody (V1NC3IDO), eFluor™ 660, eBioscience™ - FAQs (50-9750-82, 50-9750-80)