Somatostatin Monoclonal Antibody (ICDCLS), eFluor™ 660, eBioscience™ - FAQs
Somatostatin Monoclonal Antibody (ICDCLS), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Somatostatin Monoclonal Antibody (ICDCLS), eFluor™ 660, eBioscience™ - FAQs (50-9751-80, 50-9751-82)