Progesterone Receptor Monoclonal Antibody (KMC912), eFluor™ 660, eBioscience™ - FAQs
Progesterone Receptor Monoclonal Antibody (KMC912), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Progesterone Receptor Monoclonal Antibody (KMC912), eFluor™ 660, eBioscience™ - FAQs (50-9764-80)