CD4 Monoclonal Antibody (4SM95), eFluor™ 660, eBioscience™ - FAQs
CD4 Monoclonal Antibody (4SM95), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (4SM95), eFluor™ 660, eBioscience™ - FAQs (50-9766-82, 50-9766-80)