Golgin-97 Monoclonal Antibody (CDF4), eFluor™ 660, eBioscience™ - FAQs
Golgin-97 Monoclonal Antibody (CDF4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Golgin-97 Monoclonal Antibody (CDF4), eFluor™ 660, eBioscience™ - FAQs (50-9767-82, 50-9767-80)