Insulin Monoclonal Antibody (ICBTACLS), eFluor™ 660, eBioscience™ - FAQs
Insulin Monoclonal Antibody (ICBTACLS), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Insulin Monoclonal Antibody (ICBTACLS), eFluor™ 660, eBioscience™ - FAQs (50-9769-82, 50-9769-80)