SOX11 Monoclonal Antibody (SOX11-C1), eFluor™ 660, eBioscience™ - FAQs
SOX11 Monoclonal Antibody (SOX11-C1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for SOX11 Monoclonal Antibody (SOX11-C1), eFluor™ 660, eBioscience™ - FAQs (50-9773-82, 50-9773-80)