Granzyme M Monoclonal Antibody (4B2G4), eFluor™ 660, eBioscience™ - FAQs
Granzyme M Monoclonal Antibody (4B2G4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Granzyme M Monoclonal Antibody (4B2G4), eFluor™ 660, eBioscience™ - FAQs (50-9774-42, 50-9774-41)