LAMP5 (BAD-LAMP) Monoclonal Antibody (34.2), eFluor™ 660, eBioscience™ - FAQs
LAMP5 (BAD-LAMP) Monoclonal Antibody (34.2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for LAMP5 (BAD-LAMP) Monoclonal Antibody (34.2), eFluor™ 660, eBioscience™ - FAQs (50-9778-80)