Arginase 1 Monoclonal Antibody (sl6arg), eFluor™ 660, eBioscience™ - FAQs
Arginase 1 Monoclonal Antibody (sl6arg), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Arginase 1 Monoclonal Antibody (sl6arg), eFluor™ 660, eBioscience™ - FAQs (50-9779-82, 50-9779-80)