CCL8 (MCP-2) Monoclonal Antibody (DWZEE), eFluor™ 660, eBioscience™ - FAQs
CCL8 (MCP-2) Monoclonal Antibody (DWZEE), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CCL8 (MCP-2) Monoclonal Antibody (DWZEE), eFluor™ 660, eBioscience™ - FAQs (50-9789-41)