SOX2 Monoclonal Antibody (Btjce), eFluor™ 660, eBioscience™ - FAQs
SOX2 Monoclonal Antibody (Btjce), eFluor™ 660, eBioscience™ - FAQs
View additional product information for SOX2 Monoclonal Antibody (Btjce), eFluor™ 660, eBioscience™ - FAQs (50-9811-82, 50-9811-80)