GP6 Monoclonal Antibody (HY101), eFluor™ 660, eBioscience™ - FAQs
GP6 Monoclonal Antibody (HY101), eFluor™ 660, eBioscience™ - FAQs
View additional product information for GP6 Monoclonal Antibody (HY101), eFluor™ 660, eBioscience™ - FAQs (50-9813-42, 50-9813-41)