RUNX3 Monoclonal Antibody (R3-5G4), eFluor™ 660, eBioscience™ - FAQs
RUNX3 Monoclonal Antibody (R3-5G4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for RUNX3 Monoclonal Antibody (R3-5G4), eFluor™ 660, eBioscience™ - FAQs (50-9817-80, 50-9817-82)