AHR Monoclonal Antibody (FF3399), eFluor™ 660, eBioscience™ - FAQs
AHR Monoclonal Antibody (FF3399), eFluor™ 660, eBioscience™ - FAQs
View additional product information for AHR Monoclonal Antibody (FF3399), eFluor™ 660, eBioscience™ - FAQs (50-9854-42, 50-9854-41)