c-MAF Monoclonal Antibody (sym0F1), eFluor™ 660, eBioscience™ - FAQs
c-MAF Monoclonal Antibody (sym0F1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for c-MAF Monoclonal Antibody (sym0F1), eFluor™ 660, eBioscience™ - FAQs (50-9855-80, 50-9855-82)