CD369 (Clec7a, Dectin-1) Monoclonal Antibody (15E2), eFluor™ 660, eBioscience™ - FAQs
CD369 (Clec7a, Dectin-1) Monoclonal Antibody (15E2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD369 (Clec7a, Dectin-1) Monoclonal Antibody (15E2), eFluor™ 660, eBioscience™ - FAQs (50-9856-42)