IRF4 Monoclonal Antibody (3E4), eFluor™ 660, eBioscience™ - FAQs
IRF4 Monoclonal Antibody (3E4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IRF4 Monoclonal Antibody (3E4), eFluor™ 660, eBioscience™ - FAQs (50-9858-80, 50-9858-82)