BATF Monoclonal Antibody (MBM7C7), eFluor™ 660, eBioscience™ - FAQs
BATF Monoclonal Antibody (MBM7C7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for BATF Monoclonal Antibody (MBM7C7), eFluor™ 660, eBioscience™ - FAQs (50-9860-42, 50-9860-41)