Placental Alkaline Phosphatase Monoclonal Antibody (8B6), eFluor™ 660, eBioscience™ - FAQs
Placental Alkaline Phosphatase Monoclonal Antibody (8B6), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Placental Alkaline Phosphatase Monoclonal Antibody (8B6), eFluor™ 660, eBioscience™ - FAQs (50-9870-82)