Collagen IV Monoclonal Antibody (1042), eFluor™ 660, eBioscience™ - FAQs
Collagen IV Monoclonal Antibody (1042), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Collagen IV Monoclonal Antibody (1042), eFluor™ 660, eBioscience™ - FAQs (50-9871-82, 50-9871-80)