GARP Monoclonal Antibody (G14D9), eFluor™ 660, eBioscience™ - FAQs
GARP Monoclonal Antibody (G14D9), eFluor™ 660, eBioscience™ - FAQs
View additional product information for GARP Monoclonal Antibody (G14D9), eFluor™ 660, eBioscience™ - FAQs (50-9882-41, 50-9882-42)