GFAP Monoclonal Antibody (GA5), eFluor™ 660, eBioscience™ - FAQs
GFAP Monoclonal Antibody (GA5), eFluor™ 660, eBioscience™ - FAQs
View additional product information for GFAP Monoclonal Antibody (GA5), eFluor™ 660, eBioscience™ - FAQs (50-9892-80, 50-9892-82)