Vimentin Monoclonal Antibody (V9), eFluor™ 660, eBioscience™ - FAQs
Vimentin Monoclonal Antibody (V9), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Vimentin Monoclonal Antibody (V9), eFluor™ 660, eBioscience™ - FAQs (50-9897-82, 50-9897-80)