CCL5 (RANTES) Monoclonal Antibody (VL1), eFluor™ 660, eBioscience™ - FAQs
CCL5 (RANTES) Monoclonal Antibody (VL1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CCL5 (RANTES) Monoclonal Antibody (VL1), eFluor™ 660, eBioscience™ - FAQs (50-9905-42)