Gata-3 Monoclonal Antibody (TWAJ), eFluor™ 660, eBioscience™ - FAQs
Gata-3 Monoclonal Antibody (TWAJ), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Gata-3 Monoclonal Antibody (TWAJ), eFluor™ 660, eBioscience™ - FAQs (50-9966-42, 50-9966-41)