Product Image
Applied Biosystems™
ArcturusXT™ Live Cell Growth Chamber, sterile
A suite of accessories has been designed to enable research on living cells using your ARCTURUSXT™ LCM Instrument. Researchers mayRead more
Have Questions?
Catalog NumberQuantity
50003006 each
Catalog number 5000300
Price (USD)
304.00
Each
Quantity:
6 each
Price (USD)
304.00
Each
A suite of accessories has been designed to enable research on living cells using your ARCTURUSXT™ LCM Instrument. Researchers may use slides or Petri dishes to perform imaging and microdissection experiments. Following a validated protocol from ARCTURUS™ LCM Systems from Applied Biosystems™; the leaders in tissue-based microdissection – the live cells may be isolated, remaining viable for subsequent re-culture or direct processing for use in downstream applications.
Live Cell Microdissection from Petri Dishes
• Isolate living cells from a Petri Dish for subsequent re-culture
• Process collected cells for downstream molecular analysis
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Device Holds1 to 2 mL Media
For Use With (Equipment)ArcturusXT™ LCM Instrument
IncludesGrowth Chambers
Quantity6 each
Shipping ConditionRoom Temperature
Product LineARCTURUS
TypeLive Cell Growth Chamber
Unit SizeEach
Contents & Storage
Growth chambers, sterile. Store at room temperature.
Frequently asked questions (FAQs)
How do I locate the overview image on the Arcturus XT LCM Instrument?
First, optimize the visualization of the image in the live video at 2X by increasing or decreasing the brightness setting, then right-click on the overview image and select "remember settings". This resets your optimized image settings, and when you select "re-acquire overview image" you will now generate a perfectly exposed overview image.
With the ArcturusXT LCM Instrument, which laser should I use to isolate my region of interest? UV cutting laser or IR capture laser?
For a few cells: The gentle IR-only approach is the best recommendation for preserving nucleic acids and verifying that the desired material is collected on the cap. For groups of cells or large tumor regions: Mount your tissue on a membrane slide and take advantage of the power of the IR, or IR + UV cutting laser to cut around your region of interest quickly and cleanly.
What is Arcturus Laser Capture Microdissection (LCM)?
The ArcturusXT LCM Instrument has a proprietary combination of a gentle IR laser and a powerful UV laser that work in conjunction to efficiently isolate cells from frozen sections or paraffin embedded sections without changing morphology or integrity of the biological content. The IR laser helps to capture the cells of interest, and the UV laser microdissects the captured cells. This is done without affecting the morphology of the cells, and allows for visual inspection of the remaining specimen to help ensure the quality of the sample collected. In addition to the flexibility that the dual laser system provides, there is the flexibility of multiple stage inserts for various sample types. A wide-slide stage format is available for neurobiology researchers working with very large sections of brain tissue. The petri dish stage insert enables live-cell microdissection applications such as stem cell research and other rare-cell isolations.