Heraguard™ ECO Clean Bench Floor Stands
Thermo Scientific™
Heraguard™ ECO Clean Bench Floor Stands
Choose from fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Heraguard™ ECO clean bench.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
50109311Floor StandHeraguard ECO 1.8, 1.8/95
50116444Adjustable Floor StandHeraguard ECO 1.5
50051982Foot Rest BarHeraguard ECO 0.9
50051985Foot Rest BarHeraguard ECO 1.8, 1.8/95; Herasafe 2030i 1.8
50117450Electronic Adjustable Floor StandHeraguard ECO 1.5
50116441Floor StandHeraguard ECO 0.9
50117449Electronic Adjustable Floor StandHeraguard ECO 0.9
50109313Adjustable Floor StandHeraguard ECO 1.8, 1.8/95
50109312Adjustable Floor StandHeraguard ECO 1.2
50109977CastorsHeraguard ECO 0.9, 1.2, 1.5, 1.8
50051983Foot Rest BarHeraguard ECO 1.2
50109309Floor StandHeraguard ECO 1.2
50109314Electronic Adjustable Floor StandHeraguard ECO 1.2
50051984Foot Rest BarHeraguard ECO 1.5
50116442Floor StandHeraguard ECO 1.5
50116443Adjustable Floor StandHeraguard ECO 0.9
Catalog number 50109311
Price (CLP)
-
Product Type:
Floor Stand
For Use With (Equipment):
Heraguard ECO 1.8, 1.8/95
Choose from fixed, manual and electrically adjustable Thermo Scientific™ Heraguard™ ECO Clean Bench Floors Stands for the Heraguard™ ECO clean bench.
† Due to DC motors, Heraguard™ ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
DescriptionProvides ECO Work Height of 760 mm
Height Overall (Metric)680 mm
For Use With (Equipment)Heraguard ECO 1.8, 1.8/95
Product TypeFloor Stand
Unit SizeEach