Cimarec™ Biosystem 40B Controller
Actual product may vary
Thermo Scientific™
Cimarec™ Biosystem 40B Controller
Enable synchronized operation of two stirrers with Thermo Scientific™ Cimarec™ Biosystem 40B Controller.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
50118916230 V Italy
50087904115 V US
50118915230 V Germany
50118917230 V Denmark
50118918230 V UK
50118919230 V Switzerland
50118920230 V Australia
50118921230 V China
Catalog number 50118916
Price (USD)
-
Voltage:
230 V Italy
Enable synchronized operation of two stirrers with Thermo Scientific™ Cimarec™ Biosystem 40B Controller.
  • High visibility display
  • Adjustable stirring and pause times with rotation sense inversion
Specifications
DescriptionBiomodul 40B Controller 230V Italy
Electrical Requirements230 V
ModelBiomodul 40B Controller
TypeStirrer Controller (Dual)
Voltage230 V Italy
Unit SizeEach