Barnstead™ LabTower™ TII Water Purification System
Thermo Scientific™

Barnstead™ LabTower™ TII Water Purification System

Convert tap water into high-purity water with the Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System, a complete solution integrating purification and tank.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionPermeate performance
50132141Barnstead LabTower TII (UV)40 L/hr. at 15°C
50132191Barnstead LabTower TII20 L/hr. at 15°C
50132393Barnstead LabTower TII 6060 L/hr. at 15°C
Catalog number 50132141
Price (USD)
23,490.00
Online exclusive
26,100.00
Save 2,610.00
Each
Description:
Barnstead LabTower TII (UV)
Permeate performance:
40 L/hr. at 15°C
Price (USD)
23,490.00
Online exclusive
26,100.00
Save 2,610.00
Each

Convert tap water into high-purity water with the Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System, a complete solution integrating purification and tank. This high-tech system is designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity. Conserve bench space with this stand-alone design which includes an integrated 100L reservoir. Castors allow for easy relocation within the lab.

Consistent water quality:

  • Type 2 laboratory water meets ASTM Type II, CSLS-CLRW and ISO 3696 international standards.
  • The LabTower is designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity.

Compact mobile design:

  • Three system options are available with permeate flows of 20, 40 or 60L/hr.
  • Stand-alone design with integrated 100L reservoir takes up zero bench space.
  • Castors allow easy relocation.
  • Systems can be upgraded later to accommodate growing water demands.

Two ways to dispense water:

  • Dispense directly from the system via the dispensing valve for calibration solutions, reagent preparation, filling of containers, general rinsing purposes, etc.
  • Dispense from the reservoir to supply autoclaves, dishwashers, ultrapure water systems, etc.

Integrated 100L reservoir:

  • Up to 100L of high purity water is ready for dispensing when needed.
  • High purity polypropylene reservoir is opaque to light.
  • Conical bottom allows complete emptying for efficient cleaning and disinfection.
  • Adjustable setting for reservoir volume can be programmed for times when demand is low, allowing you to only store water that you can use.

Clear information on system display:

  • The large four-line display is code-protected to prevent accidental changes in system settings.
  • Illuminated and easily readable, the display provides information on resistivity or conductivity and operating mode status, such as “roduction”, “stand-by” or sanitization and volume of the reservoir in percent.

GLP-compliant documentation:

  • LabTower TII has been developed to meet or exceed GLP requirements.
  • Recorded and traceable data can be obtained by print-out via the RS-232 interface and accessory printer.
  • Highly qualified and precise measurement of the conductivity is ensured by the cell constant of 0.01 cm-1 to an accuracy of ±0.1°C.

Pretreatment:

  • External pretreatment required

Reverse osmosis and reservoir:

  • Pretreated water is pressure-forced through the permeable reverse osmosis module removing 97- 99% of all inorganic ions and 99% of dissolved organic substances, as well as microorganisms and particles.
  • The built-in 100L reservoir has a conical bottom outlet to optimize cleaning and sanitization.

Ion exchange and UV:

  • RO membrane removes up to 99% of impurities. All remaining ions are removed by the low TOC, high-purity resins in the ion exchange cartridge.
  • Irradiation with UV light (TII units with UV option only) eliminates any bacteria or germs that are present.
Specifications
Bacterial retention99%
Conductivity0.1 to 1.0μS/cm
Depth (Metric)580 mm
DescriptionBarnstead LabTower TII (UV)
Electrical Requirements100/240 V 50/60 Hz
Feedwater Pressure1 to 6 bar
Frequency50/60 Hz
Height (Metric)1500 mm
Net Weight (Metric)66 kg
Operating Pressure2 to 6 bar
Particles >0.2µM/mL<1
Permeate performance40 L/hr. at 15°C
Product TypeWater Purification System
Temperature Range (Metric)2°C to 35°C
Voltage100/240 V
Width (Metric)450 mm
Bacterial Content<1 CFU/mL
Feed Water MonitoringNo
Feedwater Connector3/4 in. NPT
Feedwater SourceTap
Flow RateLabTower TII (UV) 40 with 3 L/min. pump
Product Water TypeType 2
Resistivity10 to 1 Mohm.cm
TOC Monitor with UV Intensity MonitoringNo
Unit SizeEach