Replacement HEPA Filter
Thermo Scientific™
Replacement HEPA Filter
For Use With: Heracell VIOS 160i/250i and Forma Steri-cycle i160/i250
Have Questions?
Catalog NumberFeatures
5014192099.95% Efficiency at MPPS
Catalog number 50141920
Price (USD)
153.94
Online exclusive
179.00
Save 25.06
Each
Features:
99.95% Efficiency at MPPS
Price (USD)
153.94
Online exclusive
179.00
Save 25.06
Each
Help ensure measurable Class 10 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.95% separation efficiency @MPPS
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
Specifications
DescriptionReplacement in chamber HEPA filter, rectangular
Features99.95% Efficiency at MPPS
For Use With (Equipment)Heracell VIOS 160i/250i and Forma Steri-cycle i160/i250
LightingLED
TypeHEPA Filter
Unit SizeEach