Storage Canes
Storage Canes
Storage Canes
Storage Canes
Thermo Scientific™

Storage Canes

Store various cryostorage tubes in these storage canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDesignLength (Metric)Unit Size
5015-0002Cryo Storage CaneStandard30 cmCase of 48
378441Cryo Storage CaneStandard29 cmCase of 50
5015-0001Cryo Storage CaneStepped30 cmCase of 48
5015-0001PK
also known as 5015-0001
Cryo Storage CaneStepped30 cmPack of 12
5015-0002PK
also known as 5015-0002
Cryo Storage CaneStandard30 cmPack of 12
DS5020-0000Cryo Cane CoderCoderNot ApplicableCase of 100
Catalog number 5015-0002
Price (USD)
116.12
Online exclusive
152.25
Save 36.13
Case of 48
Type:
Cryo Storage Cane
Design:
Standard
Length (Metric):
30 cm
Unit Size:
Case of 48
Price (USD)
116.12
Online exclusive
152.25
Save 36.13
Case of 48

Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.

  • Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
  • Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
Specifications
DesignStandard
Length (English)11.8125 in.
Length (Metric)30 cm
Quantity12/Pk., 48/Cs.
MaterialAluminum
TypeCryo Storage Cane
Unit SizeCase of 48