Non-pressurized Plastic Water Deionizer Replacement Cartridges
Actual product may vary
Thermo Scientific™

Non-pressurized Plastic Water Deionizer Replacement Cartridges

These non-pressurized replacement cartridges provide deionized water for many applications and are for use with Thermo Scientific™ Non-pressurized Plastic Water Deionizer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFlow Rate
50155442100 L/hr.
5015543750 L/hr.
Catalog number 50155442
Price (USD)
-
Flow Rate:
100 L/hr.

Thermo Scientific™ Non-pressurized Plastic Water Deionizer replacement cartridges provide deionized water for many applications. For use with Non-Pressurized Plastic Water Deionizers, small compact systems that provide demineralized water. Ideal for small laboratories but also for supplying autoclaves and washers in doctors' and dentists' practices.

Specifications
DescriptionReplacement Cartridge for DI 750
Filter Size750 L
For Use With (Equipment)DI 750 non-pressurized (Part Number 03.1404)
Purity0.1 to 20 μS/cm
TypeNon-pressureized Deionizer
Flow Rate100 L/hr.
Unit SizeEach