Stainless-Steel Disposable Resin
Actual product may vary
Thermo Scientific™

Stainless-Steel Disposable Resin

Refill stainless-steel ion exchanges with this disposable, mixed-bed resin featuring advanced, regeneration service.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
50155541Resin 2 x 26 liters disposable
10.2075Resin 1 x 75 liters disposable
Catalog number 50155541
Price (USD)
-
Description:
Resin 2 x 26 liters disposable

Use Thermo Scientific™ Disposable Resin to refill stainless steel ion exchangers. The mixed-bed resins have documented quality monitoring and reliably function due to an advanced regeneration service. Each regenerated batch is monitored for both consistent capacity and quality.

This service is only available in Germany.

This service is only available in Germany.

Specifications
DescriptionResin 2 x 26 liters disposable
For Use With (Equipment)Mixed bed deionizer DI 4000 (part number 02.4000)
TypeDisposable Resin
MaterialStainless Steel
Purification MethodIon Exchange
Resin TypeMixed Bed
Unit SizeEach