Search
Search
| Catalog Number | Type | Voltage | Rotor |
|---|---|---|---|
| 50173672 | IVD-MD | 120 V | M10 Microplate (75005706), MicroClick 30 x 2 (75005719) |
| 50172635 | Multi-discipline, IVD-MD | 120 V | TX-150 (75005701), HIGHConic III (75005709), M10 Microplate (75005706), MicroClick 24 x 2 (75005715) |
| 50172637 | High Capacity Cell Culture, IVD-MD | 120 V | TX-150 (75005701) |
| 50172639 | Cell Culture, IVD-MD | 120 V | TX-150 (75005701) |
| 50172641 | Routine Clinical, IVD-MD | 120 V | TX-150 (75005701) |
| 50172645 | Molecular Biology, IVD-MD | 120 V | TX-150 (75005701), MicroClick 24 x 2 (75005715) |
| 50173671 | IVD-MD | 120 V | Microliter 48 x 2 (75003602) |