5030 SHARP Monitor
Thermo Scientific™

5030 SHARP Monitor

Take continuous high resolution measurement of airborne particulates with long and short term precision using the Thermo Scientific™ 5030 SHARP Monitor.
Have Questions?
Catalog NumberDescription
5030SHARPModel 5030 Synchronized Hybrid Ambient Real-time Particulate (SHARP) Monitor
Catalog number 5030SHARP
Price (HKD)
-
Description:
Model 5030 Synchronized Hybrid Ambient Real-time Particulate (SHARP) Monitor
Take continuous high resolution measurements of airborne particulates with long- and short-term precision using the Thermo Scientific™ 5030 SHARP Monitor. This non-stepwise hybrid ambient monitor utilizes a combination of beta attenuation and light scattering technology to measure PM-10, PM-2.5 or PM-1 mass concentrations.
The 5030 SHARP Continuous Ambient Particulate Monitor consists of a C14 source, detector and a light scattering Nephelometer in a rack-mountable enclosure. It distinguishes itself from other beta gauge measurement methods by utilizing a continuous (non-step wise) hybrid mass measurement with two proven industry technologies, which provides for long term unattended operation. It automatically advances the filter tape based upon a user defined frequency or particulate loading.

Key Features:

  • Meets U.S. and International Particulate Monitoring Regulations
  • Combines the long term accuracy of a beta gauge with the short term precision of a Nephelometer
  • Has a measurement time resolution of 1 minute (update every four seconds)
  • Simultaneously collects samples and measures on a single sample spot located between the C14 source and the proportional detector
  • Incorporates an intelligent moisture reduction (IMR) system designed to maintain the relative humidity of the air passing through the filter tape measurement assembly
  • Unit is a compact 19 in. and is rack mountable

Additional Information:

  • Dynamic digital filtering for continuous calibration update
  • Control and data exchange over two serial interfaces on monitor
  • Storage of half-hour average concentrations over a whole year
  • User-selectable reporting of mass concentration based on standard or actual flow rate
  • Calibration of all sensors is processor-controlled
  • Measurement Cycle consists of single filter spot in position for 24 hours (default); user-selectable 30 minutes to 24 hours
  • Data Averages/Values Displayed: Each full 1/2 and 24-hour values automatically stored; each full, 1/2, 1, 3, and 24-hour

Includes:

  • Temperature and humidity sensors
  • Communication cables
  • Consumables for average first year operation

Compatible with:

  • Complete Outdoor Enclosure
  • Streamline PRO Multi-Cal System

Recommended for:

  • Air quality monitoring networks, including background sites
  • Special studies and supersites
  • Routine input for air quality index
  • Remediation projects (Superfund, hazardous waste)
  • Indoor air, exposure chamber, and industrial hygiene measurements
  • In and around industrial and material handling facilities
Specifications
Flow Rate0 to 20 L/min.
Measurement MethodOptical, Silicon/Hybrid Amplifier, Radiometric, Proportional Counter (alpha and beta)
Temperature (English) Operating-22°F to +140°F
Temperature (Metric) Operating-30°C to +60°C
Accuracy±5% (compared to 24-hour FRM)
Certifications/ComplianceETL and CE compliant. U.S. EPA Equivalent PM-2.5 Monitor (EQPM-0609-184)TUV Approved
Depth (English)13 in.
Depth (Metric)4.25 cm
DescriptionModel 5030 Synchronized Hybrid Ambient Real-time Particulate (SHARP) Monitor
Frequency50/60 Hz
Height (English)12.25 in.
Height (Metric)311 cm
Measurement Range0 to 1000 μg/cu. m and 0 to 10,000 μg/cu. m (auto-ranging)
OutputsOutput: Two serial interface RS-232 (25-pin and 9-pin)Analog output: 0-10V or 4-20mA output of concentration (µg/cu.m) (specify upon order)
Precision±2μg/m3 >80μg/cu. m, ±5μg/m3 >80μg/m3, Precison between two monitors, ±0.5μg/m3
Voltage100/240 V
Weight (English)50 lb.
Weight (Metric)22.5 kg
Width (English)19 in.
Width (Metric)483 mm
Unit SizeEach