IRF7 Monoclonal Antibody (RDP4ND4), Alexa Fluor™ 647, eBioscience™ - FAQs
IRF7 Monoclonal Antibody (RDP4ND4), Alexa Fluor™ 647, eBioscience™ - FAQs
View additional product information for IRF7 Monoclonal Antibody (RDP4ND4), Alexa Fluor™ 647, eBioscience™ - FAQs (51-5375-42, 51-5375-41)