Cytomat™ 10 C450 Series Automated Incubators
Cytomat™ 10 C450 Series Automated Incubators
Actual product may vary
Thermo Scientific™

Cytomat™ 10 C450 Series Automated Incubators

Temperature Range: 4°C to 50°C with External Chiller; Relative Humidity: <98% at 37°C
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
51031168230 V
51031166100 V
51031167120 V
Catalog number 51031168
Price (MXN)
-
Voltage:
230 V

Designed to fit into the busiest of cell biology or screening laboratories, Thermo Scientific™ Cytomat™ 10 C450 wide temperature range incubator and storage unit is the ideal solution for high capacity incubation (210 SBS microplates) of primary cell or cell line cultures with a wide temperature application range combined with 4°C storage. The latest product of the Cytomat 10 series can be seamless integrated with existing automated assay hardware and provides mean access time below 10 seconds combined with controlled humidity and thermal decontamination routine ContraCon.

Matches your Capacity with 10 Stacker
  • 280 pcs. 1536-well plates
  • 210 pcs. 96-well or 384-well plates
  • 150 pcs. transwell or insert plates
  • 100 pcs. deep-well

–Further stacker types on request


Effortless Humidity Control
  • HydraSmart Technology - Ultrasonic vapor technology
  • Sterilized water supply with external tank

Fast
  • Below 10 sec. mean access time

Contamination Prevention
  • Automated decontamination routine - ContraCon
  • Solid copper inner chamber (option)
  • H2O2-flange (option)

Convenient – Easy to use
  • 5" Touch screen as standard
  • Various gate / transfer positions

More options
  • O2 control
  • high Humidity (hH) up to 98% r.H. at 37°C
  • Internal and direct barcode reader

Certification
  • CE
  • RoHS III

Manufactured in ISO 9001 and ISO 13485 accredited facility

Specifications
Access TimeBelow 10 sec.
Certifications/ComplianceCE Marked
Depth (English)32.3 in.
Depth (Metric)822 mm
Electrical Requirements230 V, 50 Hz
FeaturesIncubation and storage unit with temperature range below with external chiller, humidity up to 95% r.H. at 37°C and CO2 range of 0-20 Vol. % at 25°C to 50°C
Height (English)36.0 in.
Height (Metric)915 mm
Holds280 Pc. 1536-Well Plates, 210 Pc. 96-Well or 384-Well Plates, 150 Pc. Transwell or Insert Plates, 100 Pc. Deep-Well
TypeAutomated Incubator
Width (English)31.5 in.
Width (Metric)800 mm
Chamber MaterialCopper
Relative Humidity<95%
Temperature Range (Metric)4°C to 50°C (with External Chiller)
Voltage230 V
Unit SizeEach