Cytomat™ 5 C Series Automated Incubators
Actual product may vary
Thermo Scientific™

Cytomat™ 5 C Series Automated Incubators

Temperature Range: Ambient +10°C to 50°C; Relative Humidity: <98% at 37°C
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
51031526100 V
51031527120 V
51031528230 V
Catalog number 51031526
Price (MXN)
-
Voltage:
100 V

Designed to fit into the busiest cell biology or screening laboratories, the Thermo Scientific™ Cytomat™ 5 C Series Automated Incubator is ideal for midsize capacity (105 SBS microplates) cell growth and assay incubation. Cytomat 5 C Series incubators have a temperature range of ambient +10°C to 50°C combined with controlled humidity up to 98% r.H., stable CO2 conditions, and a thermal decontamination routine. They seamlessly integrate with existing automated assay hardware, provide mean access time below 10 seconds, and significantly decrease the risk of contamination and reduce assay variability.

5-Stacker to Match Your Capacity
  • 140 pcs. 1536-well plates
  • 105 pcs. 96-well or 384-well plates
  • 75 pcs. transwell or insert plates
  • 50 pcs. deep-well

Mix and match different pitch stackers. Additional stacker types are available upon request.

Effortless Humidity Control
  • HydraSmart technology provides ultrasonic vapor technology with no turbulence and heat introduction into system
  • Sterilized water supply by external accessible micro-particle filter
  • Closed loop water delivery system, with simple external tank
  • Water level monitoring with refill warning
  • High humidity up to 98% r.H. at 37°C (optional)
Fast
  • Below 10 second access time
  • Application speed settings: slow, medium, fast
  • Individual microplate speed settings
Contamination Prevention
  • ContraCon for an automated decontamination routine with true walk-away time
  • Easy to clean - smooth stainless steel surface
  • Solid copper inner chamber (optional)
  • H2O2-flange (optional)
Flexible Stacker Design
  • Compatible with existing Cytomat stacker
  • Standard or customized stacker pitch sizes
  • Mix and match different pitch stackers
  • Autoclaveable stainless steel
  • Ventilation slots allow accurate temperature distribution
Convenient, Easy to Use
  • 5-inch touch screen with LED backlight as standard
  • Various gate / transfer positions
Easy to Integrate
  • Intuitive software commands
  • Flat gate, no gate cover
  • Designed with multiple transfer stations
More Options
  • Additional capacity: one additional stacker position on turntable
  • O2 control
  • Solid copper inner chamber
  • H2O2 flange
  • Internal and direct barcode reader
Certification
  • CE
  • RoHS III
  • Manufactured in ISO 9001 and ISO 13485 accredited facility
Specifications
Access TimeBelow 10 sec.
Certifications/ComplianceCE Marked
Depth (English)32.3 in.
Depth (Metric)822 mm
Electrical Requirements100 V, 50 Hz
FeaturesIncubation and Storage Unit with Temperature Range Below, Humidity up to 98% r.H. (at 37°C and CO2 Range of 0-20 Vol. %)
Height (English)36.0 in.
Height (Metric)915 mm
Holds140 Pc. 1536-Well Plates, 105 Pc. 96-Well or 384-Well Plates, 75 Pc. Transwell or Insert Plates, 50 Pc. Deep-Well
TypeAutomated Incubator
Width (English)31.5 in.
Width (Metric)800 mm
Chamber MaterialStainless Steel
MaterialStainless Steel
Relative Humidity<98%
Temperature Range (Metric)Ambient +10° to 50°C
Voltage100 V
Unit SizeEach