Multiskan™ FC Microplate Photometer (EU IVD/CE-marked) - FAQs
Multiskan™ FC Microplate Photometer (EU IVD/CE-marked) - FAQs
View additional product information for Multiskan™ FC Microplate Photometer (EU IVD/CE-marked) - FAQs (51119010)
24 product FAQs found
Is Multiskan FC integrateable with robotics?
Yes, the Multiskan FC is intergrateable with robotics.
My Multiskan FC needs a new lamp. What is the catalog or part number? Can I replace it myself?
Yes, the Halogen lamp 6V/10W is Cat. No. 1410101. You can replace the lamp yourself using instructions in the Multiskan FC User Manual: https://www.thermofisher.com/order/catalog/product/51119100
I think my Multiskan FC isn't working right, but there's no error message. How can I check?
If the results are not as expected, performance can be verified using Multiskan Verification Plate (Cat. Nos. 24072800 (for 8 wavelengths, use with either model) or 24072805 (for 16 wavelengths, use with 51119100)).
Do I need to calibrate my Multiskan FC?
Readers are factory-calibrated and perform self-diagnostics on start-up to ensure consistent results over time. Should an error message appear indicating calibration is needed, contact your local Thermo Fisher Scientific microplate instrument service group at thermofisher.com/contactus.
Can I use uDrop Plate with Multiskan FC?
uDrop Plate is not compatible with Multiskan FC. For UV applications, refer to Multiskan Sky: https://www.thermofisher.com/order/catalog/product/51119500
Can I read my DNA / RNA / protein at 260 / 280 in Multiskan FC?
No, Multiskan FC reads only in the visible range, 340-800 nm. For UV applications, refer to Multiskan Sky: https://www.thermofisher.com/order/catalog/product/51119500
Do I have to use a PC with software to read plates with Multiskan FC?
A computer is not required. The instrument has on-board control that allows for programming and analysis of most assays.
Does the Multiskan FC include software with purchase?
Yes, SkanIt Research Edition is included with purchase; SkanIt Drug Discovery Edition for meeting CFR 21 Part 11 regulations (Cat. No. 5187149) must be purchased separately.
Does the Multiskan FC have a touchscreen?
No, it has a Graphical User Interface with keypad for navigation.
What are the dimensions / weight of Multiskan FC?
The dimensions and weight of the Mjultiskan FC are: ca. 29 cm (W) x 40 cm (D) x 22 cm (H); weight 8.5 kg.
Can I put Multiskan FC in a cold room?
The cold room must meet these operating conditions: +10C to +40C; maximum relative humidity 80% for temperatures up to 31 degrees C decreasing linearly to 50% relative humidity at 40 degrees C, non-condensing; indoor use only.
What are the electrical specifications for Multiskan FC?
The electrical specifications are as follows: 100-240 Vac, 50/60 Hz, nominal.
Is Multiskan FC CE / cSGSus / CSA / NRTL / UL / RoHS certified?
Multiskan FC carries the following certifications: CE, cSGSus, CSA, NRTL, UL, RoHS.
Which plate should I use with Multiskan FC?
We recommend using SBS-standard flat-bottom plates of clear polystyrene that have a total height no taller than 15.3 mm (including any lid). Cat. No. 51119000 can accept 96 well plates; Cat. No. 51119100 can accept either 96 or 384 well plates. U bottom plates are acceptable but not ideal; V bottom plates cannot be used. No specific part number is required as long as it meets the above specifications.
Does the Multiskan FC include shaking?
Yes, both models have linear shaking standard.
Does the Multiskan FC have incubation? What is the range?
The Multiskan FC With Incubator (Cat. No. 51119100) has incubation from ambient +4 to 50 degrees C. The Multiskan FC (Cat. No. 51119000) does not have incubation.
Can the Multiskan FC read 6 / 12 / 24 / 48 well plates?
The Multiskan FC Cat. No. 51119000 can read 96 well plates; the Multiskan FC Cat. No. 51119100 can read 96- and 384-well plates.
Can the Multiskan FC read 384 well plates?
The Multiskan FC With Incubator (Cat. No. 51119100) can read 384-well plates. The Multiskan FC (Cat. No. 51119000) reads only 96-well plates.
What is the linearity of Multiskan FC?
At 405 nm: 0-3 Abs, +/- 2%, 96-well plate; 0-2.5 Abs, +/- 2%, 384-well plate.
How long does it take Multiskan FC to read my plate?
Read speed / measurement speed in Fast mode:
96-well plate = 7 seconds
384-well plate = 13 seconds
What is the half-bandwidth for Multiskan FC filters?
The HBW for a filter will range from 3-9 nm.
How many filters does Multiskan FC hold?
The filter wheel holds up to 8 filters.
What is the part number for additional Multiskan FC filters?
A complete list of available filters with part numbers can be found in the Multiskan FC User Manual which can be found on this page: https://www.thermofisher.com/order/catalog/product/51119100
What filters ship with Multiskan FC?
405, 450 and 650 nm ship with every Multiskan FC. No substitutions are allowed. A complete list of available filters for separate purchase can be found in the Multiskan FC User Manual found on the following page: https://www.thermofisher.com/order/catalog/product/51119100