Multiskan™ FC 96 and 384-well Plate Microplate Photometers
Thermo Scientific™

Multiskan™ FC 96 and 384-well Plate Microplate Photometers

The Multiskan FC Microplate Photometer is a reliable instrument for a wide variety of research and routine applications. It reads 96- and 384-well plates and is equipped with shaking and incubation capabilities for temperature critical assays. It can be used as a stand-alone instrument or under PC control with intuitive Thermo Scientific SkanIt software.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypePlate Type
51119100Multiskan FC with Incubator96 and 384-well Plates
51119000Multiskan FC96-well Plate
Catalog number 51119100
Price (USD)
11,990.00
Each
Product Type:
Multiskan FC with Incubator
Plate Type:
96 and 384-well Plates
Price (USD)
11,990.00
Each

Multiskan FC brings together over 40 years experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard. A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.

Features:

  • Proven performance and reliable day-to-day results through patented optical design and in-built self-diagnostics
  • A broad wavelength range of 340–850 nm for a wide variety of research and routine applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves
  • Equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 nm and 620 nm, preinstalled
  • A comprehensive range of easy-to install additional filters available for order
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • Shaking and incubation up to 50°C for temperature critical assays
  • Ease of use through the large color screen and a variety of language versions
  • Visual and logical SkanIt software for comprehensive instrument control and data handling
  • Multiskan verification plate available for instrument performance verification
  • Robot compatibility for high-throughput environments
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
  • USB port for easy data transfer

Recommended for:

Immunoassays (ELISA), protein assays, endotoxins, cytotoxicity and proliferation assays, enzyme assays, growth curves

For Research Use Only.
Specifications
Accuracy± 1% (0 to 3 Abs) or ± 0.003 Abs, which ever is greater at 405 nm
CompatibilityRobot
Depth (English)15.7 in.
Depth (Metric)40 cm
DescriptionMultiskan FC with incubator
Filters8-position filter wheel; filters installed: 405 nm, 450 nm, and 620 nm
Height (English)8.7 in.
Height (Metric)22 cm
Hertz50/60 Hz
Incubator TemperatureAmbient +4° to 50°C
Linearity96-well plate: 0 to 3 Abs, ±2% at 405 nm, 384-well plate: 0 to 2.5 Abs, ±2% at 405 nm
Measurement Speed7 sec. for 96-well Plate (Fast Mode), 13 sec. for 96-well Plate (Normal Mode)
PrecisionCV ≤ 0.2% (0.3 to 3 Abs) at 405 nm
Product TypeMultiskan FC with Incubator
Resolution0.001 Abs
Software TypeSkanIt Software
Voltage100/240 V
Warranty1 Year
Wavelength Range340 to 850 nm
Width (English)11.4 in.
Width (Metric)29 cm
Detector TypePhotodetector
InterfaceOn-board or PC Control
Light SourceQuartz Halogen Lamp
Measurement TechnologyAbsorbance
Plate Type96 and 384-well Plates
Product LineMultiskan
ShakingLinear
Wavelength SelectionFilters
Unit SizeEach

Frequently asked questions (FAQs)

Can I calibrate the Multiskan FC Microplate Photometer (Fisherbrand accuSkan FC Filter-Based Microplate Photometer)?

The Multiskan FC Microplate Photometer (same instrument as the Fisherbrand accuSkan FC Filter-Based Microplate Photometer) is factory-calibrated and performs self-diagnostics on start-up to ensure consistent results over time. If you need to verify that the instrument is working properly, one of the options below can be utilized:

- Performance can be user-verified using Multiskan Verification Plate (Cat. No. 24072800, for 8 wavelengths or Cat. No. 24072805, for 16 wavelengths).

- The other option for verification is by purchasing our qualification service as an IQ/OQ/PQ or IPV that is required for laboratories that operate within a quality management system, comply with global standards (e.g., GLP, GCP, GMP, ISO 17025, and ISO 15189), and/or follow country-specific regulations (e.g., US FDA 21 CFR Part 11, CLIA, and CAP). Our qualification service provides documented verification that laboratory instruments are installed and operating according to the manufacturer’s performance specifications. More information about this service can be found here.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What other filters are available for the Multiskan FC Microplate Photometer in addition to the three standard filters (405, 450, and 620 nm)?

The Multiskan FC Microplate Photometer offers filters with the following wavelengths for separate purchase. The filter SKU is 142xxx5, where xxx = filter wavelength.
340 nm
375 nm
414 nm
492 nm
520 nm
530 nm
540 nm
550 nm
560 nm
570 nm
595 nm
630 nm
650 nm
690 nm
740 nm
750 nm
820 nm

Is Multiskan FC integrateable with robotics?

Yes, the Multiskan FC is intergrateable with robotics.

My Multiskan FC needs a new lamp. What is the catalog or part number? Can I replace it myself?

Yes, the Halogen lamp 6V/10W is Cat. No. 1410101. You can replace the lamp yourself using instructions in the Multiskan FC User Manual: https://www.thermofisher.com/order/catalog/product/51119100

I think my Multiskan FC isn't working right, but there's no error message. How can I check?

If the results are not as expected, performance can be verified using Multiskan Verification Plate (Cat. Nos. 24072800 (for 8 wavelengths, use with either model) or 24072805 (for 16 wavelengths, use with 51119100)).