Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Thermo Scientific™

Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature RangeArrayHolds
5116-16000°C3 x 412 Tubes (16 to 17 mm dia.)
355501-20°C4 x 520 Tubes (0.2 to 0.5 mL)
5115-0012-20°C3 x 412 Tubes (0.2 to 0.5 mL)
5115-0032-20°C4 x 832 Tubes (0.2 to 0.5 mL)
5116-13000°C3 x 412 Tubes (12 to 13 mm dia.)
DS5116-00120°C3 x 412 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
DS5116-00320°C4 x 832 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
DS5114-0012-20°C3 x 412 Cryogenic Vials (0.2 to 1.5 mL)
Catalog number 5116-1600
Price (USD)
253.65
Online exclusive
313.64
Save 59.99
Each
Temperature Range:
0°C
Array:
3 x 4
Holds:
12 Tubes (16 to 17 mm dia.)
Price (USD)
253.65
Online exclusive
313.64
Save 59.99
Each

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications
Array3 x 4
Description3x4 compartments, for 16-17mm dia.vials
DesignNon-filled/clear lid
Dimensions (L x W x H)7.75 x 5.5 x 7.5 in. (197 x 140 x 190 mm)
Height (English)7.5 in.
Height (Metric)190 mm
Holds Temp., Hours<1°C, 5.0
Length (English)7.75 in.
Length (Metric)197 mm
Width (English)5.5 in.
Width (Metric)140 mm
Capacity (Metric)0.2 to 0.5 mL
ColorClear
Holds12 Tubes (16 to 17 mm dia.)
MaterialPolycarbonate
Product TypeBenchtop Cooler
Temperature Range0°C
Unit SizeEach