Sterilin™ Autoclave Bags
Sterilin™ Autoclave Bags
Sterilin™ Autoclave Bags
Sterilin™ Autoclave Bags
Thermo Scientific™

Sterilin™ Autoclave Bags

Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)MaterialNo. per Case
511HT810 x 610mmPolypropylene200
509660 x 305mmHDPE200
509L610 x 406mmHDPE200
510-LCD660 x 305mmHDPE500
510L610 x 406mmHDPE500
511810 x 610mmHDPE200
510HT660 x 305mmPolypropylene500
509LHT610 x 406mmPolypropylene200
510LHT601 x 406mmPolypropylene500
Catalog number 511HT
Price (USD)
-
Dimensions (L x W):
810 x 610mm
Material:
Polypropylene
No. per Case:
200
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags

Ordering Alerts:

Not available in all geographic regions

Specifications
DescriptionAutoclave bags (PP)
Dimensions (L x W)810 x 610mm
Length (Metric)810 mm
No. per Case200
Temperature Range (Metric)135°C
Width (Metric)610 mm
Length (English)32 in.
MaterialPolypropylene
Width (English)24 in.
Unit SizeCase of 200